Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose | 57-48-7 | 50 G
SDP

Catalog No. 5000418967
Encompass_Preferred

D-Fructose, also known as D(-)-Fructose, is a monosaccharide found naturally in sources like honey, fruits, and various plants. It functions as an endogenous metabolite.

  • Molecular weight: 180.16
  • Formula: C6H12O6
  • Appearance: Solid, white to off-white
  • Purity: ≥98.0%
  • Combines to form the disaccharide sucrose
  • Absorbed by the small intestine and processed by the liver
  • For research use only
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Plants, Gramineae, Saccharum officinarum
  • Endogenous metabolite
  • Shipping: Room temperature
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-18967 Supplier Medchemexpress LLC Supplier No. HYN709250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose, also known as D(-)-Fructose, is a monosaccharide found naturally in sources like honey, fruits, and various plants. It functions as an endogenous metabolite.

  • Molecular weight: 180.16
  • Formula: C6H12O6
  • Appearance: Solid, white to off-white
  • Purity: ≥98.0%
  • Combines to form the disaccharide sucrose
  • Absorbed by the small intestine and processed by the liver
  • For research use only
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Plants, Gramineae, Saccharum officinarum
  • Endogenous metabolite
  • Shipping: Room temperature
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.