Learn More
Medchemexpress LLC D-fructose-6-phosphate-13C6 (disodium x.hydrate) | 00-00-0 | 99.9% | 13C6H4Na2O9P·XH2O | 5 MG
SDP
Catalog No. 5000222699
Encompass_Preferred
D-Fructose-6-phosphate-13C6 disodium x.hydrate is a stable isotope-labeled form of D-fructose-6-phosphate provided as the disodium salt in hydrated form. It is intended for use as an internal standard or tracer in liquid chromatography-mass spectrometry and metabolic flux analysis, where uniform 13C labeling enables accurate quantitation and tracing of metabolic pathways.
  • Uniform 13C6 labeling for accurate isotope tracing.
  • High purity (99.9%) suitable for analytical workflows.
  • Disodium salt in hydrated form for solubility and handling.
  • Available in small pack sizes appropriate for internal standard use.
Catalog No. 50-002-22699 Supplier Medchemexpress LLC Supplier No. HY144085S5MG
D-Fructose-6-phosphate-13C6 disodium x.hydrate is a stable isotope-labeled form of D-fructose-6-phosphate provided as the disodium salt in hydrated form. It is intended for use as an internal standard or tracer in liquid chromatography-mass spectrometry and metabolic flux analysis, where uniform 13C labeling enables accurate quantitation and tracing of metabolic pathways.
  • Uniform 13C6 labeling for accurate isotope tracing.
  • High purity (99.9%) suitable for analytical workflows.
  • Disodium salt in hydrated form for solubility and handling.
  • Available in small pack sizes appropriate for internal standard use.