Learn More
Medchemexpress LLC D-Fructose-6-phosphate-13C6 disodium x.hydrate | 643-13-0 | 99.90% | 13C6H4Na2O9P.XH2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY144085S1MG
Encompass_Preferred
D-Fructose-6-phosphate-13C6 disodium x.hydrate is a 13C-labeled D-Fructose-6-phosphate disodium x.hydrate, intended for research use only. This compound serves as a valuable tool in various scientific applications, particularly as a tracer and an internal standard for quantitative analysis. Its stable heavy isotopes of carbon are incorporated into the molecule, allowing for precise tracking and measurement in research settings. This characteristic is especially useful in drug development processes to study pharmacokinetics and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes for research applications.
Catalog No. 50-003-55430