Learn More
AdipoGen D-Fructose 6-phosphate 2Na H2O
SDP
Catalog No. 502560864
Encompass_Preferred
Chemical. CAS 26177-86-6. Formula C6H11O9P . 2Na. MW 304.1. Synthetic. A glycolytic intermediate formed by the isomerization of glucose-6-phosphate.
Catalog No. 50-256-0864 Supplier AdipoGen Supplier No. CDXF0212M500
Chemical. CAS 26177-86-6. Formula C6H11O9P . 2Na. MW 304.1. Synthetic. A glycolytic intermediate formed by the isomerization of glucose-6-phosphate.