Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-fructose-6-phosphate | 98.0% | 260.14 g·mol⁻¹ | C6H13O9P | 5 MG
SDP

Catalog No. 5000263932
Encompass_Preferred

D-fructose-6-phosphate is an endogenous sugar phosphate involved in glycolysis and the pentose phosphate pathway. Supplied as a solid research standard, it is suitable for enzymatic assays, metabolomics, and pathway studies. The free form can be prone to instability; handle sealed and protect from moisture.

  • Solid, white to off-white powder.
  • Molecular formula C6H13O9P; molecular weight 260.14 g·mol⁻¹.
  • Typically 98.0% purity suitable for analytical and enzymatic assays.
  • Recommended storage refrigerated; in solution store at -80°C for long-term stability.
  • Used as a metabolic intermediate standard in biochemical research.

Catalog No. 50-002-63932 Supplier Medchemexpress LLC Supplier No. HY113407CR5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-fructose-6-phosphate is an endogenous sugar phosphate involved in glycolysis and the pentose phosphate pathway. Supplied as a solid research standard, it is suitable for enzymatic assays, metabolomics, and pathway studies. The free form can be prone to instability; handle sealed and protect from moisture.

  • Solid, white to off-white powder.
  • Molecular formula C6H13O9P; molecular weight 260.14 g·mol⁻¹.
  • Typically 98.0% purity suitable for analytical and enzymatic assays.
  • Recommended storage refrigerated; in solution store at -80°C for long-term stability.
  • Used as a metabolic intermediate standard in biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.