Learn More
Medchemexpress LLC D-Fructose-6-phosphate disodium | 26177-86-6 | 99.9% | 304.10 | 10 MG
SDP
Catalog No. 5000437906
Encompass_Preferred
D-Fructose 6-phosphate disodium is an endogenous metabolite. It can be obtained by hydrolysis of Fructose-1,6-bisphosphatase (FBPase). It is a sugar intermediate in the glycolysis pathway and the pentose phosphate pathway. D-Fructose-6-phosphate disodium can be used to detect glucosamine-6-phosphate synthase and TAL activities. D-Fructose 6-phosphate can also be used to study Lewy body dementia.
  • Endogenous metabolite
  • Can be obtained by hydrolysis of Fructose-1,6-bisphosphatase (FBPase)
  • Sugar intermediate in the glycolysis pathway and the pentose phosphate pathway
  • Can be used to detect glucosamine-6-phosphate synthase and TAL activities
  • Can be used to study Lewy body dementia
  • For research use only
Catalog No. 50-004-37906 Supplier Medchemexpress LLC Supplier No. HY113407A10MG
D-Fructose 6-phosphate disodium is an endogenous metabolite. It can be obtained by hydrolysis of Fructose-1,6-bisphosphatase (FBPase). It is a sugar intermediate in the glycolysis pathway and the pentose phosphate pathway. D-Fructose-6-phosphate disodium can be used to detect glucosamine-6-phosphate synthase and TAL activities. D-Fructose 6-phosphate can also be used to study Lewy body dementia.
  • Endogenous metabolite
  • Can be obtained by hydrolysis of Fructose-1,6-bisphosphatase (FBPase)
  • Sugar intermediate in the glycolysis pathway and the pentose phosphate pathway
  • Can be used to detect glucosamine-6-phosphate synthase and TAL activities
  • Can be used to study Lewy body dementia
  • For research use only