Learn More
Medchemexpress LLC D-Fructose-6-phosphate disodium | 26177-86-6 | 99.9% | 304.10 | 25 MG
SDP
Catalog No. 5000437631
Encompass_Preferred
D-Fructose-6-phosphate disodium is an endogenous metabolite that can be obtained by the hydrolysis of Fructose-1,6-bisphosphatase (FBPase). It is a sugar intermediate involved in both the glycolysis pathway and the pentose phosphate pathway. This product can be utilized for detecting glucosamine-6-phosphate synthase and transaldolase (TAL) activities. Furthermore, D-Fructose 6-phosphate disodium can be used in studies related to Lewy body dementia.
  • Naturally occurring within biological systems.
  • Functions as a sugar intermediate in glycolysis and the pentose phosphate pathway.
  • Can be used to detect glucosamine-6-phosphate synthase and TAL activities.
  • Useful for studying conditions like Lewy body dementia.
Catalog No. 50-004-37631 Supplier Medchemexpress LLC Supplier No. 25MGHY113407CR
D-Fructose-6-phosphate disodium is an endogenous metabolite that can be obtained by the hydrolysis of Fructose-1,6-bisphosphatase (FBPase). It is a sugar intermediate involved in both the glycolysis pathway and the pentose phosphate pathway. This product can be utilized for detecting glucosamine-6-phosphate synthase and transaldolase (TAL) activities. Furthermore, D-Fructose 6-phosphate disodium can be used in studies related to Lewy body dementia.
  • Naturally occurring within biological systems.
  • Functions as a sugar intermediate in glycolysis and the pentose phosphate pathway.
  • Can be used to detect glucosamine-6-phosphate synthase and TAL activities.
  • Useful for studying conditions like Lewy body dementia.