Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-fructose 6-phosphate disodium | 26177-86-6 | 99.9% | 304.10 g/mol | C6H11Na2O9P | 5 MG
SDP

Catalog No. 5000264180
Encompass_Preferred

D-fructose 6-phosphate disodium is the disodium salt of the glycolytic intermediate fructose-6-phosphate, supplied as a high-purity solid research reagent (CAS 26177-86-6). It is typically used as an analytical standard, enzyme substrate, and research tool in metabolic and biochemical assays.

  • High purity solid form suitable for biochemical assays.
  • CAS 26177-86-6 and molecular weight 304.10 g/mol for precise identification.
  • Used as substrate or standard in enzymatic and metabolic pathway studies.
  • Store at -20°C in a sealed container to protect from moisture.
  • Available in small analytical sizes including 5 mg for research applications.

Catalog No. 50-002-64180 Supplier Medchemexpress LLC Supplier No. HY113407A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-fructose 6-phosphate disodium is the disodium salt of the glycolytic intermediate fructose-6-phosphate, supplied as a high-purity solid research reagent (CAS 26177-86-6). It is typically used as an analytical standard, enzyme substrate, and research tool in metabolic and biochemical assays.

  • High purity solid form suitable for biochemical assays.
  • CAS 26177-86-6 and molecular weight 304.10 g/mol for precise identification.
  • Used as substrate or standard in enzymatic and metabolic pathway studies.
  • Store at -20°C in a sealed container to protect from moisture.
  • Available in small analytical sizes including 5 mg for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.