Learn More
Medchemexpress LLC D-Fructose-6-phosphate disodium monohydrate | 99.88% | 322.11 | 1 MG
SDP
Catalog No. 5000435713
Encompass_Preferred
D-Fructose-6-phosphate disodium monohydrate (Standard) is the analytical standard of D-Fructose-6-phosphate disodium monohydrate. D-Fructose-6-phosphate is an intermediate of Glycolysis and a precursor for the synthesis of amino sugars, such as galactosamine and glucosamine. It is for research use only and not sold to patients.
  • Is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Is an intermediate of glycolysis and a precursor for the synthesis of amino sugars.
  • For research use only.
Catalog No. 50-004-35713 Supplier Medchemexpress LLC Supplier No. HY113407CR1MG
D-Fructose-6-phosphate disodium monohydrate (Standard) is the analytical standard of D-Fructose-6-phosphate disodium monohydrate. D-Fructose-6-phosphate is an intermediate of Glycolysis and a precursor for the synthesis of amino sugars, such as galactosamine and glucosamine. It is for research use only and not sold to patients.
  • Is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Is an intermediate of glycolysis and a precursor for the synthesis of amino sugars.
  • For research use only.