Learn More
Medchemexpress LLC D-Fructose-d | 374089-83-5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S8100MG
Encompass_Preferred
D-Fructose-d is a deuterium-labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. This stable heavy isotope of D-Fructose is primarily used in research and analytical applications where precise tracking and quantitation of metabolic processes are essential. Deuterium labeling offers advantages in studying drug metabolism and pharmacokinetics due to its potential to alter metabolic profiles.
  • Deuterium labeled D-Fructose for metabolic research
  • Suitable as a tracer in various biological studies
  • Can be used as an internal standard for quantitative analysis
  • Compatible with analytical techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-17851