Learn More
Alkali Scientific D Fructose [Levulose], 5 Kg
SDP
Supplier:  Alkali Scientific DFR003CP
Encompass_Preferred
D-Fructose (levulose) is a laboratory-grade sugar used extensively in biochemical research. This 5kg supply is particularly useful for high-throughput applications, including cell culture studies and metabolic research. Fructose serves as a carbon source in microbiological assays and metabolic studies, providing energy for microbial and mammalian cells. It is frequently used in experiments focused on insulin resistance, obesity, diabetes, and metabolic disorders. In addition, fructose is incorporated into culture media for the study of enzyme activity and carbohydrate metabolism, making it an essential reagent for many biochemical and pharmaceutical research projects.
Catalog No. 50-006-60930