Learn More
Alkali Scientific D Fructose [Levulose], 500 G
SDP
Supplier:  Alkali Scientific DFR002CP
Encompass_Preferred
D-Fructose (levulose) is a simple sugar used in a wide range of biochemical research applications. This 500g laboratory-grade quantity is essential for studying carbohydrate metabolism and its role in energy production. D-Fructose is commonly used as a carbon source in microbial and mammalian cell cultures, enabling the study of metabolic processes. It is also used in the formulation of growth media and in experiments exploring insulin resistance, diabetes, and other metabolic diseases. As a versatile reagent, fructose plays a key role in various research fields, particularly those related to metabolism and endocrinology.
Catalog No. 50-006-59786