Learn More
Medchemexpress LLC D-Fructose (Standard) | 57-48-7 | 25 MG
SDP
Catalog No. 5000421819
Encompass_Preferred
D-Fructose (Standard) is the analytical standard of D-Fructose, a monosaccharide found in honey, fruits, and other plants. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. With a molecular weight of 180.16 and a chemical formula of C6H12O6, this compound appears as a white to off-white solid with an assay of 99.62%. When ingested orally, D-Fructose is absorbed by the small intestine and flows through the liver, resulting in a higher concentration in the liver than in the blood vessels.
  • Analytical standard for D-Fructose
  • Intended for research and analytical applications
  • Monosaccharide found in honey, fruits, and plants
  • Used in HPLC, GC, and MS experiments
  • Appears as a white to off-white solid
  • Assay of 99.62%
Catalog No. 50-004-21819 Supplier Medchemexpress LLC Supplier No. 25MGHYN7092R
D-Fructose (Standard) is the analytical standard of D-Fructose, a monosaccharide found in honey, fruits, and other plants. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. With a molecular weight of 180.16 and a chemical formula of C6H12O6, this compound appears as a white to off-white solid with an assay of 99.62%. When ingested orally, D-Fructose is absorbed by the small intestine and flows through the liver, resulting in a higher concentration in the liver than in the blood vessels.
  • Analytical standard for D-Fructose
  • Intended for research and analytical applications
  • Monosaccharide found in honey, fruits, and plants
  • Used in HPLC, GC, and MS experiments
  • Appears as a white to off-white solid
  • Assay of 99.62%