Learn More
Medchemexpress LLC D-fructose (Standard) | 57-48-7 | MFCD00148910 | 500 MG
SDP
Catalog No. 5000420162
Encompass_Preferred
D-Fructose (Standard) is an analytical standard of D-Fructose, a monosaccharide found in honey, fruits, and other plants. This product is intended for research and analytical applications.
  • Analytical standard for D-Fructose
  • Suitable for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS techniques
Catalog No. 50-004-20162 Supplier Medchemexpress LLC Supplier No. HYN7092R500MG
D-Fructose (Standard) is an analytical standard of D-Fructose, a monosaccharide found in honey, fruits, and other plants. This product is intended for research and analytical applications.
  • Analytical standard for D-Fructose
  • Suitable for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS techniques