Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Galactal 98.0+%, TCI America™
SDP

Catalog No. G02731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02731G Supplier TCI America Supplier No. G02731G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21193-75-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00038067
InChI Key YVECGMZCTULTIS-HSUXUTPPSA-N
Synonym d-galactal, galactal, d-+-galactal, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol, 2r,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4-diol, arabino-hex-5-enitol, 2,6-anhydro-5-deoxy, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, pubchem10609
PubChem CID 2734735
ChEBI CHEBI:63139
IUPAC Name (2R,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
SMILES C1=COC(C(C1O)O)CO

Specifications

Quantity 1 g
Formula Weight 146.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-Galactal
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.