Learn More
Medchemexpress LLC D-galacto-D-mannan | 11078-30-1 | MFCD00146683 | 95.0% | ~200,000 g/mol | (C6H10O5)n | 25 MG
SDP
Catalog No. 5000283001
Encompass_Preferred
D-Galacto-D-mannan (carob galactomannan) is a nonionic, linear polysaccharide used as a high-molecular-weight hydrocolloid for thickening, stabilizing, co-gelling, and fat replacement in formulation and analytical applications. Typical preparations are purified and suitable for research and formulation studies involving polysaccharide rheology and stabilization.
Catalog No. 50-002-83001 Supplier Medchemexpress LLC Supplier No. HYW145481A25MG
D-Galacto-D-mannan (carob galactomannan) is a nonionic, linear polysaccharide used as a high-molecular-weight hydrocolloid for thickening, stabilizing, co-gelling, and fat replacement in formulation and analytical applications. Typical preparations are purified and suitable for research and formulation studies involving polysaccharide rheology and stabilization.