Learn More
Medchemexpress LLC D-galacto-D-mannan | 11078-30-1 | MFCD00146683 | 95.0% | 504.4 g/mol | C18H32O16 | 1 MG
SDP
Catalog No. 5000283821
Encompass_Preferred
D-Galacto-D-mannan is a plant-derived galactomannan polysaccharide isolated from Ceratonia siliqua (carob). Supplied for research use at approximately 95% purity, it serves as a reference reagent and assay component for carbohydrate analysis, biochemical studies, and immunological applications.
  • Plant-derived galactomannan from Ceratonia siliqua.
  • Approximately 95.0% purity by HPLC for analytical use.
  • Suitable as a reference standard for carbohydrate composition analysis.
  • Useful as a reagent in biochemical and immunological assays.
  • Available in small, precisely measured quantities for research applications.
  • Polymeric material with molecular weight dependent on degree of polymerization.
Catalog No. 50-002-83821 Supplier Medchemexpress LLC Supplier No. HYW1454811MG
D-Galacto-D-mannan is a plant-derived galactomannan polysaccharide isolated from Ceratonia siliqua (carob). Supplied for research use at approximately 95% purity, it serves as a reference reagent and assay component for carbohydrate analysis, biochemical studies, and immunological applications.
  • Plant-derived galactomannan from Ceratonia siliqua.
  • Approximately 95.0% purity by HPLC for analytical use.
  • Suitable as a reference standard for carbohydrate composition analysis.
  • Useful as a reagent in biochemical and immunological assays.
  • Available in small, precisely measured quantities for research applications.
  • Polymeric material with molecular weight dependent on degree of polymerization.