Learn More
Medchemexpress LLC D-galacto-D-mannan | 11078-30-1 | MFCD00146683 | 95.0% | 504.4 g/mol | C18H32O16 | 50 MG
SDP
Catalog No. 5000283206
Encompass_Preferred
D-Galacto-D-mannan (carob galactomannan) is a nonionic, linear polysaccharide extracted from Ceratonia siliqua. It is commonly used in research and formulation work as a thickener, stabilizer, co-gellant, and fat replacer, and is supplied as a white to off-white solid with defined purity and storage recommendations.
  • Nonionic, linear polysaccharide derived from Ceratonia siliqua.
  • Functions as a thickener, stabilizer, co-gellant, and fat replacer.
  • Provided as a white to off-white solid suitable for formulation studies.
  • High purity (95.0%) for consistent research results.
  • Stable when stored as powder at -20°C or 4°C for recommended durations.
Catalog No. 50-002-83206 Supplier Medchemexpress LLC Supplier No. HYW145481A50MG
D-Galacto-D-mannan (carob galactomannan) is a nonionic, linear polysaccharide extracted from Ceratonia siliqua. It is commonly used in research and formulation work as a thickener, stabilizer, co-gellant, and fat replacer, and is supplied as a white to off-white solid with defined purity and storage recommendations.
  • Nonionic, linear polysaccharide derived from Ceratonia siliqua.
  • Functions as a thickener, stabilizer, co-gellant, and fat replacer.
  • Provided as a white to off-white solid suitable for formulation studies.
  • High purity (95.0%) for consistent research results.
  • Stable when stored as powder at -20°C or 4°C for recommended durations.