Learn More
Medchemexpress LLC D-Galacto-D-mannan (carob galactomannan) | 11078-30-1 | MFCD00146683 | 95.0% | Not specified (polymeric; variable molecular weight) | C18H32O16 | 100 MG
SDP
Catalog No. 5000283042
Encompass_Preferred
D-Galacto-D-mannan is a carob-derived (Ceratonia siliqua) linear, nonionic polysaccharide used in biochemical applications as a thickener, stabilizer, co-gellant, and fat replacer. The material is supplied as a white powder for research use only and is typically stored refrigerated or frozen per handling guidance.
Catalog No. 50-002-83042 Supplier Medchemexpress LLC Supplier No. HYW145481A100MG
D-Galacto-D-mannan is a carob-derived (Ceratonia siliqua) linear, nonionic polysaccharide used in biochemical applications as a thickener, stabilizer, co-gellant, and fat replacer. The material is supplied as a white powder for research use only and is typically stored refrigerated or frozen per handling guidance.