Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-D-galactopyranose-1-13C, 1-(dihydrogen phosphate), dipotassium salt | 478518-78-4 | MFCD08459758 | 98.8% | 337.31 g/mol | C6H11K2O9P (1-13C) | 5 MG
SDP

Catalog No. 5000264286
Encompass_Preferred

Galactose 1-phosphate-13C potassium is a stable-isotope labeled potassium salt of galactose 1-phosphate supplied as a high-purity, white to off-white solid for research use. It is used as a labeled internal standard and as an intermediate for studies of galactose metabolism and nucleotide-sugar pathways. Available in small milligram sizes for analytical and method development workflows.

  • Stable-isotope labeled internal standard for quantitative mass spectrometry.
  • Intermediate in galactose metabolism and nucleotide-sugar pathways.
  • High purity (98.8%) suitable for analytical applications.
  • White to off-white solid, convenient for handling and storage.
  • Available in small milligram package sizes for method development.
  • Molecular weight 337.31 g/mol; formula C6H11K2O9 with one 13C label.

Catalog No. 50-002-64286 Supplier Medchemexpress LLC Supplier No. HY113143AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Galactose 1-phosphate-13C potassium is a stable-isotope labeled potassium salt of galactose 1-phosphate supplied as a high-purity, white to off-white solid for research use. It is used as a labeled internal standard and as an intermediate for studies of galactose metabolism and nucleotide-sugar pathways. Available in small milligram sizes for analytical and method development workflows.

  • Stable-isotope labeled internal standard for quantitative mass spectrometry.
  • Intermediate in galactose metabolism and nucleotide-sugar pathways.
  • High purity (98.8%) suitable for analytical applications.
  • White to off-white solid, convenient for handling and storage.
  • Available in small milligram package sizes for method development.
  • Molecular weight 337.31 g/mol; formula C6H11K2O9 with one 13C label.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.