Learn More
Medchemexpress LLC β-D-galactopyranose, 1,2,3,4,6-pentaacetate | 4163-60-4 | MFCD00063259 | 98.0% | 390.34 g·mol⁻¹ | C16H22O11 | 500 G
SDP
Catalog No. 5000301257
Encompass_Preferred
β-D-Galactose pentaacetate is an acetylated derivative of D-galactose used as a research reagent. It has been reported to act as a negative regulator of insulin, penetrates plasma membranes, and is hydrolyzed by intracellular esterases. Provided for laboratory research use in solid and solution formats, with safety and quality documents available.
Catalog No. 50-003-01257 Supplier Medchemexpress LLC Supplier No. HYW039923500G
β-D-Galactose pentaacetate is an acetylated derivative of D-galactose used as a research reagent. It has been reported to act as a negative regulator of insulin, penetrates plasma membranes, and is hydrolyzed by intracellular esterases. Provided for laboratory research use in solid and solution formats, with safety and quality documents available.