Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-Galactopyranose, 1,2,3,4,6-pentaacetate | 4163-60-4 | MFCD00063259 | 99.6% | 390.34 g·mol⁻¹ | C16H22O11 | 100 G
SDP

Catalog No. 5000300214
Encompass_Preferred

β-D-Galactopyranose, 1,2,3,4,6-pentaacetate is the per-O-acetylated derivative of D-galactopyranose. It is supplied as a white to off-white solid used as a protected sugar building block and a cell-permeable biochemical tool that is hydrolyzed by intracellular esterases to release the parent sugar; common uses include studies of insulin regulation and carbohydrate synthesis.

  • High purity: 99.6% assay.
  • Molecular weight 390.34 g·mol⁻¹.
  • Chemical formula C16H22O11.
  • Soluble in DMSO (100 mg/mL) and methanol (16.67 mg/mL).
  • Store at 4°C under nitrogen, away from moisture.
  • Available in gram-scale solid quantities and solution formats for convenient use.

Catalog No. 50-003-00214 Supplier Medchemexpress LLC Supplier No. HYW039923100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Galactopyranose, 1,2,3,4,6-pentaacetate is the per-O-acetylated derivative of D-galactopyranose. It is supplied as a white to off-white solid used as a protected sugar building block and a cell-permeable biochemical tool that is hydrolyzed by intracellular esterases to release the parent sugar; common uses include studies of insulin regulation and carbohydrate synthesis.

  • High purity: 99.6% assay.
  • Molecular weight 390.34 g·mol⁻¹.
  • Chemical formula C16H22O11.
  • Soluble in DMSO (100 mg/mL) and methanol (16.67 mg/mL).
  • Store at 4°C under nitrogen, away from moisture.
  • Available in gram-scale solid quantities and solution formats for convenient use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.