Learn More
Medchemexpress LLC D-galactopyranose, 2-azido-2-deoxy-, 1,3,4,6-tetraacetate | 84278-00-2 | MFCD02683262 | >97.0% | 373.32 g/mol | C14H19N3O9 | 250 MG
SDP
Catalog No. 5000284659
Encompass_Preferred
D-Galactopyranose, 2-azido-2-deoxy-, 1,3,4,6-tetraacetate is an azido-functional, tetra-acetylated D-galactopyranose reagent used in glycobiology and carbohydrate chemistry. It is employed as a protected 2-azido-2-deoxy galactose derivative for synthetic modifications, analytical workflows, and biochemical assay preparation.
  • Azido functional group suitable for click chemistry and further derivatization.
  • Tetra-acetyl protection improves stability and solubility in organic solvents.
  • Applicable to glycobiology research and carbohydrate synthesis.
  • Molecular formula C14H19N3O9; molecular weight 373.32 g/mol.
  • Reported purity ≥97% by HPLC (supplier data).
Catalog No. 50-002-84659 Supplier Medchemexpress LLC Supplier No. HYW686970250MG
D-Galactopyranose, 2-azido-2-deoxy-, 1,3,4,6-tetraacetate is an azido-functional, tetra-acetylated D-galactopyranose reagent used in glycobiology and carbohydrate chemistry. It is employed as a protected 2-azido-2-deoxy galactose derivative for synthetic modifications, analytical workflows, and biochemical assay preparation.
  • Azido functional group suitable for click chemistry and further derivatization.
  • Tetra-acetyl protection improves stability and solubility in organic solvents.
  • Applicable to glycobiology research and carbohydrate synthesis.
  • Molecular formula C14H19N3O9; molecular weight 373.32 g/mol.
  • Reported purity ≥97% by HPLC (supplier data).