Learn More
Medchemexpress LLC D-galactopyranose, 2-azido-2-deoxy-, 1,3,4,6-tetraacetate | 84278-00-2 | MFCD02683262 | 98.0% | 373.32 g/mol | C14H19N3O9 | 500 MG
SDP
Catalog No. 5000284567
Encompass_Preferred
D-Galactopyranose, 2-azido-2-deoxy-, 1,3,4,6-tetraacetate is a tetra-O-acetylated azido-sugar derivative supplied as a research reagent (CAS 84278-00-2). It serves as a protected glycosyl building block and introduces an azide handle for downstream functionalization.
  • Protected azido-sugar building block for synthetic chemistry.
  • Enables azide-based click chemistry (CuAAC) derivatization.
  • Suitable for glycosylation and conjugation workflows.
  • Supplied as a solid, typical package 500 MG.
  • Molecular formula C14H19N3O9; molecular weight 373.32 g/mol.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84567 Supplier Medchemexpress LLC Supplier No. HYW686970500MG
D-Galactopyranose, 2-azido-2-deoxy-, 1,3,4,6-tetraacetate is a tetra-O-acetylated azido-sugar derivative supplied as a research reagent (CAS 84278-00-2). It serves as a protected glycosyl building block and introduces an azide handle for downstream functionalization.
  • Protected azido-sugar building block for synthetic chemistry.
  • Enables azide-based click chemistry (CuAAC) derivatization.
  • Suitable for glycosylation and conjugation workflows.
  • Supplied as a solid, typical package 500 MG.
  • Molecular formula C14H19N3O9; molecular weight 373.32 g/mol.
  • For research use only; not for human or clinical use.