Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Α-D-Galactopyranoside, 1,6-dichloro-1,6-dideoxy-β-D-fructofuranosyl 4-chloro-4-deoxy- | 56038-13-2 | 397.63 | 5 G
SDP

Catalog No. 5000389091
Encompass_Preferred

Sucralose is a non-nutritive artificial sweetener and sugar substitute. It can activate a conserved neural fasting response, leading to an appetite-stimulating effect in rodents. It is for research use only.

  • Non-nutritive artificial sweetener
  • Sugar substitute
  • Activates conserved neural fasting response
  • Purity: 98.0%
  • Appearance: White to off-white solid

Catalog No. 50-003-89091 Supplier Medchemexpress LLC Supplier No. HYN06145G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sucralose is a non-nutritive artificial sweetener and sugar substitute. It can activate a conserved neural fasting response, leading to an appetite-stimulating effect in rodents. It is for research use only.

  • Non-nutritive artificial sweetener
  • Sugar substitute
  • Activates conserved neural fasting response
  • Purity: 98.0%
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.