Learn More
Medchemexpress LLC Β-D-Galactopyranoside, 4-aminophenyl | 5094-33-7 | 99.5% | 271.27 | 50 MG
SDP
Catalog No. 5000452572
Encompass_Preferred
4-Aminophenyl-β-D-galactopyranoside, 98% is a biochemical reagent. It serves as a substrate for β-galactosidase.
  • Biochemical reagent
  • Substrate for β-galactosidase
Catalog No. 50-004-52572 Supplier Medchemexpress LLC Supplier No. HYW01165450MG
4-Aminophenyl-β-D-galactopyranoside, 98% is a biochemical reagent. It serves as a substrate for β-galactosidase.
  • Biochemical reagent
  • Substrate for β-galactosidase