Learn More
Medchemexpress LLC D(+)-Galactosamine hydrochloride (Standard) | 1772-03-8 | 99.95% | 215.63 | 50 MG
SDP
Catalog No. 5000403953
Encompass_Preferred
D(+)-Galactosamine (hydrochloride) (Standard) is the analytical standard for D(+)-Galactosamine (hydrochloride), intended for research and analytical applications. It is an established experimental toxin that primarily causes liver injury through the generation of free radicals and depletion of UTP nucleotides. Intoxication with D(+)-Galactosamine hydrochloride also induces renal dysfunction, with renal failure often accompanying the end-stage of liver damage. The Lipopolysaccharide/D(+)-Galactosamine-induced acute liver injury model is a known animal model used for fulminant hepatic failure.
  • Analytical standard for D(+)-Galactosamine (hydrochloride).
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • The compound is a grade of analytical standard, serving as a reference standard for assay.
  • Available in various quantities (e.g., 10 mg, 25 mg, 50 mg, 100 mg, 250 mg).
Catalog No. 50-004-03953 Supplier Medchemexpress LLC Supplier No. HY42682R50MG
D(+)-Galactosamine (hydrochloride) (Standard) is the analytical standard for D(+)-Galactosamine (hydrochloride), intended for research and analytical applications. It is an established experimental toxin that primarily causes liver injury through the generation of free radicals and depletion of UTP nucleotides. Intoxication with D(+)-Galactosamine hydrochloride also induces renal dysfunction, with renal failure often accompanying the end-stage of liver damage. The Lipopolysaccharide/D(+)-Galactosamine-induced acute liver injury model is a known animal model used for fulminant hepatic failure.
  • Analytical standard for D(+)-Galactosamine (hydrochloride).
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • The compound is a grade of analytical standard, serving as a reference standard for assay.
  • Available in various quantities (e.g., 10 mg, 25 mg, 50 mg, 100 mg, 250 mg).