Learn More
Medchemexpress LLC D-Galactosamine pent 100g | 76375-60-5 | 100 G
SDP
Catalog No. 5000226389
Encompass_Preferred
D-galactosamine pentaacetate is the penta-acetylated derivative of D-galactosamine supplied as a solid research reagent. It is used as a protected amino-sugar building block and intermediate in carbohydrate and glycoconjugate synthesis, and is appropriate for organic synthesis and analytical applications.
  • Acetylated derivative of D-galactosamine for use as a protected amino-sugar reagent.
  • Common intermediate in carbohydrate and glycan synthesis.
  • Supplied as a solid suitable for laboratory research.
  • Molecular formula C16H23NO10 and molecular weight 389.35 g/mol.
  • Available in multiple package sizes, including 100 G.
Catalog No. 50-002-26389 Supplier Medchemexpress LLC Supplier No. HY117271100G
D-galactosamine pentaacetate is the penta-acetylated derivative of D-galactosamine supplied as a solid research reagent. It is used as a protected amino-sugar building block and intermediate in carbohydrate and glycoconjugate synthesis, and is appropriate for organic synthesis and analytical applications.
  • Acetylated derivative of D-galactosamine for use as a protected amino-sugar reagent.
  • Common intermediate in carbohydrate and glycan synthesis.
  • Supplied as a solid suitable for laboratory research.
  • Molecular formula C16H23NO10 and molecular weight 389.35 g/mol.
  • Available in multiple package sizes, including 100 G.