Learn More
Medchemexpress LLC D-Galactosamine pent 1g | 76375-60-5 | 389.35 g/mol | C16H23NO10 | 1 G
SDP
Catalog No. 5000228772
Encompass_Preferred
D-Galactosamine pentaacetate is an acetylated derivative of D-galactosamine provided as a solid reagent for life-science research and organic synthesis. The compound is characterized by a molecular formula C16H23NO10, molecular weight ~389.35 g/mol, and batch-reported purity ≥98% (NMR). It is commonly used in carbohydrate chemistry, glycosylation studies, and as a synthetic intermediate.
  • Acetylated derivative suitable for synthetic chemistry
  • High purity (≥98% NMR) for analytical and preparative use
  • Solid form, easy to weigh and handle
  • Documented SDS and COA available for batch-specific data
  • Suitable as a building block in carbohydrate modification and glycosylation studies
Catalog No. 50-002-28772 Supplier Medchemexpress LLC Supplier No. HY1172711G
D-Galactosamine pentaacetate is an acetylated derivative of D-galactosamine provided as a solid reagent for life-science research and organic synthesis. The compound is characterized by a molecular formula C16H23NO10, molecular weight ~389.35 g/mol, and batch-reported purity ≥98% (NMR). It is commonly used in carbohydrate chemistry, glycosylation studies, and as a synthetic intermediate.
  • Acetylated derivative suitable for synthetic chemistry
  • High purity (≥98% NMR) for analytical and preparative use
  • Solid form, easy to weigh and handle
  • Documented SDS and COA available for batch-specific data
  • Suitable as a building block in carbohydrate modification and glycosylation studies