Learn More
Medchemexpress LLC D-Galactosamine pent 25g | 76375-60-5 | C16H23NO10 | 25 G
SDP
Catalog No. 5000226790
Encompass_Preferred
D-Galactosamine pentaacetate is an acetyl-protected derivative of D-galactosamine (CAS 76375-60-5) used as a biochemical reagent and synthetic intermediate in life-science research. The solid compound has formula C16H23NO10 and molecular weight 389.35 g·mol-1.
  • Used as a biochemical reagent and synthetic intermediate.
  • Acetyl-protected amino sugar for selective transformations.
  • Molecular weight 389.35 g·mol-1, formula C16H23NO10.
  • Provided as a 25 G pack for laboratory use.
  • Intended for research use only and not for human or diagnostic use.
Catalog No. 50-002-26790 Supplier Medchemexpress LLC Supplier No. HY11727125G
D-Galactosamine pentaacetate is an acetyl-protected derivative of D-galactosamine (CAS 76375-60-5) used as a biochemical reagent and synthetic intermediate in life-science research. The solid compound has formula C16H23NO10 and molecular weight 389.35 g·mol-1.
  • Used as a biochemical reagent and synthetic intermediate.
  • Acetyl-protected amino sugar for selective transformations.
  • Molecular weight 389.35 g·mol-1, formula C16H23NO10.
  • Provided as a 25 G pack for laboratory use.
  • Intended for research use only and not for human or diagnostic use.