Learn More
Medchemexpress LLC D-Galactosamine pent 5g | 76375-60-5 | 5 G
SDP
Catalog No. 5000228545
Encompass_Preferred
D-galactosamine pentaacetate is the per-acetylated derivative of D-galactosamine supplied as a laboratory reagent for use in carbohydrate chemistry and synthetic organic workflows. It serves as a protected building block for glycosylation, derivatization, and analytical studies.
  • Per-acetylated D-galactosamine derivative suitable for synthesis.
  • High purity (≥98.0%) confirmed by certificate of analysis.
  • Molecular weight 389.35 g/mol for formulation and calculation.
  • Useful as a protected building block in carbohydrate chemistry.
  • Provided with SDS and COA for safe handling and traceability.
Catalog No. 50-002-28545 Supplier Medchemexpress LLC Supplier No. HY1172715G
D-galactosamine pentaacetate is the per-acetylated derivative of D-galactosamine supplied as a laboratory reagent for use in carbohydrate chemistry and synthetic organic workflows. It serves as a protected building block for glycosylation, derivatization, and analytical studies.
  • Per-acetylated D-galactosamine derivative suitable for synthesis.
  • High purity (≥98.0%) confirmed by certificate of analysis.
  • Molecular weight 389.35 g/mol for formulation and calculation.
  • Useful as a protected building block in carbohydrate chemistry.
  • Provided with SDS and COA for safe handling and traceability.