Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-Galactosamine pentaacetate | 3006-60-8 | 389.35 | 10 G
SDP

Catalog No. 5000449338
Encompass_Preferred

β-D-Galactosamine pentaacetate is an intermediate utilized in the synthesis of triantennary N-Acetylgalactosamine ligands. It is also suitable for research into liver-targeted delivery of siRNA.

  • Intermediate in the synthesis of triantennary N-Acetylgalactosamine ligands
  • Can be studied in liver-targeted delivery of siRNA research
  • Appears as a white to off-white solid
  • Stable under recommended storage conditions
  • For research use only
  • Molecular formula: C16H23NO10
  • Melting/freezing point: 234-236°C
  • Boiling point/range: 530.2±50.0 °C at 760 mmHg

Catalog No. 50-004-49338 Supplier Medchemexpress LLC Supplier No. HYW00572910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Galactosamine pentaacetate is an intermediate utilized in the synthesis of triantennary N-Acetylgalactosamine ligands. It is also suitable for research into liver-targeted delivery of siRNA.

  • Intermediate in the synthesis of triantennary N-Acetylgalactosamine ligands
  • Can be studied in liver-targeted delivery of siRNA research
  • Appears as a white to off-white solid
  • Stable under recommended storage conditions
  • For research use only
  • Molecular formula: C16H23NO10
  • Melting/freezing point: 234-236°C
  • Boiling point/range: 530.2±50.0 °C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.