Learn More
Cayman Chemical D-Galactosamine (hydrochloride), Purity: ≥98%, CAS: 1772-03-8, 25 g
SDP
Catalog No. 502994573
Encompass_Preferred
A hepatotoxic agent; an amino sugar derivative of galactose; used, alone or in combination with LPS, as a model of liver failure in rodents
Catalog No. 50-299-4573 Supplier Cayman Chemical Supplier No. 2298125 G
A hepatotoxic agent; an amino sugar derivative of galactose; used, alone or in combination with LPS, as a model of liver failure in rodents