Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals alpha-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%

Catalog No. AC337030025
Change view
Click to view available options
Quantity:
50 mg
500 mg
2.5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC337030025 2.5 g Glass bottle
AC337030500 50 mg Glass bottle
AC337035000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC337030025 Supplier Thermo Scientific Chemicals Supplier No. 337030025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19046-60-7 | C6H11K2O9P | 336.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19046-60-7
Molecular Formula C6H11K2O9P
Molecular Weight (g/mol) 336.32
MDL Number MFCD00065475
InChI Key KCIDZIIHRGYJAE-YGFYJFDDSA-L
Synonym alpha-d-galactose-1-phosphate dipotassium salt dihydrate
PubChem CID 131875454
IUPAC Name potassium;[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
SMILES [K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Color Colorless to White
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 g
Assay Percent Range 98%
Solubility Information (5% in water) Clear colorless
Formula Weight 426.39
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Appearance (5% aq. soln.) clear, colorless
Chemical Name or Material α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 242-783-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.