Learn More
Alkali Scientific D (+) Galactose, 100 G
SDP
Supplier:  Alkali Scientific DGA001CP
Encompass_Preferred
D (+) Galactose, 100 G, is a high-purity carbohydrate reagent used for various biochemical and analytical applications. This D-form galactose is essential in laboratory settings for studying sugar metabolism, enzymatic reactions, and glycosylation processes. It is commonly used as a substrate in cell culture media, microbiology, and genetic research. Galactose is vital in metabolic studies, especially in assays measuring the activity of galactose-related enzymes such as galactokinase. In addition, it is used in biochemical studies for testing the properties of galactose-utilizing microorganisms and as a growth supplement in certain media formulations. The 100g package is ideal for smaller-scale applications.
Catalog No. 50-006-60389