Learn More
Selleck Chemical LLC D-Galactose S3849-1g
SDP
Catalog No. 503109013
Encompass_Preferred
D-Galactose (D-Galactopyranose D-( )-Galactose) is an aldohexose that occurs naturally in the D-form in lactose cerebrosides gangliosides and mucoproteins and is converted enzymatically into D-glucose for metabolism or polysaccharides for storage It accelerates senescence in invertebrates and mammals and has been used as a model for aging D-Galactose can be used to induce animal models of Senescence
Catalog No. 50-310-9013 Supplier Selleck Chemical LLC Supplier No. S38491G
D-Galactose (D-Galactopyranose D-( )-Galactose) is an aldohexose that occurs naturally in the D-form in lactose cerebrosides gangliosides and mucoproteins and is converted enzymatically into D-glucose for metabolism or polysaccharides for storage It accelerates senescence in invertebrates and mammals and has been used as a model for aging D-Galactose can be used to induce animal models of Senescence