Learn More
Selleck Chemical LLC D-Galactose-100mg
SDP
Supplier:  Selleck Chemical LLC S3849100MG
Encompass_Preferred
D-Galactose (D-Galactopyranose, D-(+)-Galactose) is an aldohexose that occurs naturally in the D-form in lactose, cerebrosides, gangliosides, and mucoproteins and is converted enzymatically into D-glucose for metabolism or polysaccharides for storage. It accelerates senescence in invertebrates and mammals and has been used as a model for aging.
Catalog No. 50-238-9990