Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(+)-galactose-13C-2 | 478518-56-8 | 99.9% | 181.15 g/mol | C5^13CH12O6 | 10 MG
SDP

Catalog No. 5000296286
Encompass_Preferred

D-Galactose-13C-2 is D-galactose labeled with a single 13C isotope at the C-2 position. It is supplied as a stable isotope-labeled standard for metabolic tracing, isotope dilution quantification, and mass spectrometry applications; for research use only.

  • 13C label at C-2 for metabolic tracing.
  • High chemical purity: 99.9%.
  • Molecular weight 181.15 g/mol.
  • CAS number 478518-56-8.
  • Supplied as a 10 mg vial suitable for analytical use.
  • Stable isotope standard for quantitative and qualitative assays.

Catalog No. 50-002-96286 Supplier Medchemexpress LLC Supplier No. HYN0210S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Galactose-13C-2 is D-galactose labeled with a single 13C isotope at the C-2 position. It is supplied as a stable isotope-labeled standard for metabolic tracing, isotope dilution quantification, and mass spectrometry applications; for research use only.

  • 13C label at C-2 for metabolic tracing.
  • High chemical purity: 99.9%.
  • Molecular weight 181.15 g/mol.
  • CAS number 478518-56-8.
  • Supplied as a 10 mg vial suitable for analytical use.
  • Stable isotope standard for quantitative and qualitative assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.