Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-galactose-13C6 | 74134-89-7 | MFCD01075538 | 99.0% | 186.11 g/mol | 13C6H12O6 | 10 MG
SDP

Catalog No. 5000295670
Encompass_Preferred

D-Galactose-13C6 is the 13C6 isotopically labeled form of D-galactose supplied for research use. It serves as a stable-isotope tracer and internal standard for quantitative analysis in metabolic and analytical studies by NMR, GC-MS, and LC-MS, and is the C-4 epimer of glucose.

  • High isotopic and analytical purity (99.0%).
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Available in small analytical sizes for tracer studies.
  • Stable powder with recommended storage: powder -20°C (long term) or 4°C (short term).
  • Molecular formula: 13C6H12O6; molecular weight: 186.11 g/mol.

Catalog No. 50-002-95670 Supplier Medchemexpress LLC Supplier No. HYN0210S1610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Galactose-13C6 is the 13C6 isotopically labeled form of D-galactose supplied for research use. It serves as a stable-isotope tracer and internal standard for quantitative analysis in metabolic and analytical studies by NMR, GC-MS, and LC-MS, and is the C-4 epimer of glucose.

  • High isotopic and analytical purity (99.0%).
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Available in small analytical sizes for tracer studies.
  • Stable powder with recommended storage: powder -20°C (long term) or 4°C (short term).
  • Molecular formula: 13C6H12O6; molecular weight: 186.11 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.