Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-galactose, 2-amino-2-deoxy-, hydrochloride (1:1) | 1772-03-8 | MFCD00135830 | 100.0% | 215.63 g/mol | C6H14ClNO5 | 10 MG
SDP

Catalog No. 5000297876
Encompass_Preferred

D(+)-Galactosamine hydrochloride is an analytical reference standard supplied as a high-purity solid hydrochloride salt for research and analytical applications. It is used in toxicology and mechanistic studies and to induce experimental hepatic injury in animal models (e.g., LPS/D-galactosamine models), and is suitable for small-scale analytical assays.

  • CAS number 1772-03-8.
  • Molecular formula C6H14ClNO5.
  • Molecular weight 215.63 g/mol.
  • Appearance: solid.
  • Form: hydrochloride salt.
  • Purity: 99.95% (as listed).
  • Available in small sizes including 10 mg for analytical use.

Catalog No. 50-002-97876 Supplier Medchemexpress LLC Supplier No. HY42682R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D(+)-Galactosamine hydrochloride is an analytical reference standard supplied as a high-purity solid hydrochloride salt for research and analytical applications. It is used in toxicology and mechanistic studies and to induce experimental hepatic injury in animal models (e.g., LPS/D-galactosamine models), and is suitable for small-scale analytical assays.

  • CAS number 1772-03-8.
  • Molecular formula C6H14ClNO5.
  • Molecular weight 215.63 g/mol.
  • Appearance: solid.
  • Form: hydrochloride salt.
  • Purity: 99.95% (as listed).
  • Available in small sizes including 10 mg for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.