Learn More
Bioworld D(+) Galactose, 500 g
Catalog No. 50256017
Encompass_Preferred
An monosaccharidic aldo hexose potentially useful in aging studies. Specifications:High Purity GradeSpecific Rotation (2% water): 79 - 81°Appearance: White PowderBoiling Point : 410.8 °CDensity : 1.5 g/cm3HS Code : 29400090Melting Point: 168-170 °CRTECS Number: LW5490000
Catalog No. 50-256-017 Supplier Bioworld Supplier No. 407000003
An monosaccharidic aldo hexose potentially useful in aging studies. Specifications:High Purity GradeSpecific Rotation (2% water): 79 - 81°Appearance: White PowderBoiling Point : 410.8 °CDensity : 1.5 g/cm3HS Code : 29400090Melting Point: 168-170 °CRTECS Number: LW5490000