Learn More
Gold Biotechnology Inc D Galactose 500 g
SDP
Supplier:  Gold Biotechnology Inc G010500
Encompass_Preferred
D-Galactose is a monosaccharide sugar that serves as an energy source and glycosylation component It is a C-4 epimer of glucose and often used as a source of carbon in culture media Galactose is a component of the disaccharide lactose and released upon hydrolysis by -galactosidase enzymes It is converted to glucose via the Leloir pathway or metabolized via an alternative pathway such as the DeLey-Doudoroff pathway
Catalog No. 50-005-62157