Learn More
Medchemexpress LLC D-galactose | 59-23-4 | MFCD00151230 | 100.0% | 180.16 g/mol | C6H12O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0210R10MG
Encompass_Preferred
D-Galactose (Standard) is an analytical reference standard of D-galactose supplied as a high-purity crystalline material for use in qualitative and quantitative laboratory assays. It is intended as a calibration and reference material for analytical methods and is provided with supporting documentation such as a certificate of analysis and SDS.
  • High purity for accurate calibration and quantitation.
  • Suitable for HPLC, GC, and MS analytical techniques.
  • Provided with a certificate of analysis for traceability.
  • Available in small pack sizes for analytical workflows.
Catalog No. 50-002-96356