Learn More
Medchemexpress LLC D-galactose-6-O-sulfate sodium salt | 125455-62-1 | MFCD00078771 | 95.0% | 282.20 g·mol⁻¹ | C6H11NaO9S | 10 MG
SDP
Catalog No. 5000282921
Encompass_Preferred
D-Galactose-6-O-sulfate sodium salt is a chemically defined carbohydrate reagent used in life-science research to study glycan sulfation, carbohydrate-protein interactions, and enzymatic assays. Supplied as the sodium salt, it is provided as a solid suitable for analytical and biochemical applications.
  • Chemically defined monosaccharide sulfated at the 6-O position.
  • Suitable for glycan and carbohydrate-protein interaction studies.
  • High purity, typically 95.0%.
  • Molecular weight 282.20 g·mol⁻¹ and formula C6H11NaO9S.
  • Store sealed, away from moisture at -20°C; follow recommended storage for solutions.
  • Available in small research pack sizes for analytical assays.
Catalog No. 50-002-82921 Supplier Medchemexpress LLC Supplier No. HYW14567310MG
D-Galactose-6-O-sulfate sodium salt is a chemically defined carbohydrate reagent used in life-science research to study glycan sulfation, carbohydrate-protein interactions, and enzymatic assays. Supplied as the sodium salt, it is provided as a solid suitable for analytical and biochemical applications.
  • Chemically defined monosaccharide sulfated at the 6-O position.
  • Suitable for glycan and carbohydrate-protein interaction studies.
  • High purity, typically 95.0%.
  • Molecular weight 282.20 g·mol⁻¹ and formula C6H11NaO9S.
  • Store sealed, away from moisture at -20°C; follow recommended storage for solutions.
  • Available in small research pack sizes for analytical assays.