Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™

Catalog No. ICN19467080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19467080 100 g
ICN19467090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19467080 Supplier MP Biomedicals, Inc Supplier No. 0219467080
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10257-28-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym d-galactose, d-galactopyranose, galactose, galactopyranose, galactopyranoside, d-+-galactose, galactose, pure, cerebrose, gal, d-galactopyranoside
PubChem CID 6036
ChEBI CHEBI:4139

Specifications

Quantity 100 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.