Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Galactose Anhydrous, Fisher Chemical

Catalog No. G1100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
G1-100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G1-100

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00151230
Synonym Cerebrose, Brain Sugar

Specifications

Alpha Vector Galactose Anhydrous
Melting Point 167°C
Color White
Packaging Glass Bottle
Quantity 100 g
Formula Weight 180.16
Grade Reagent
Physical Form Solid
Chemical Name or Material D-(+)-Galactose Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.