Learn More
Medchemexpress LLC D-Galactose pentaace 5g
SDP
Catalog No. 5000206949
Encompass_Preferred
D-Galactose pentaacetate inhibits leucine-induced insulin release D-Galactose pentaacetate can be used for the research of persistent hyperinsulinemia in childhood or insulinoma[1][2]
Catalog No. 50-002-06949 Supplier Medchemexpress LLC Supplier No. HYW1023875G
D-Galactose pentaacetate inhibits leucine-induced insulin release D-Galactose pentaacetate can be used for the research of persistent hyperinsulinemia in childhood or insulinoma[1][2]