Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-galactose pentaacetate | 4163-60-4 | MFCD00063259 | 99.6% | 390.34 g·mol-1 | C16H22O11 | 250 G
SDP

Catalog No. 5000303617
Encompass_Preferred

β-D-galactose pentaacetate is the β-anomer of D-galactose with five acetylated hydroxyl groups. It functions as an insulin antagonist, penetrates the plasma membrane, and is hydrolyzed by intracellular esterases; it inhibits leucine-induced insulin release in Wistar rat islets. Supplied as a solid or as pre-made DMSO solutions for research use.

  • High purity (≈99.6%).
  • Molecular weight 390.34 g·mol-1.
  • Cell-permeable acetylated sugar cleaved by intracellular esterases.
  • Available as solid and ready-to-use DMSO solutions.
  • White to off-white solid appearance.
  • Store solid at 4°C under nitrogen and protect from moisture.

Catalog No. 50-003-03617 Supplier Medchemexpress LLC Supplier No. 250GHYW039923
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-galactose pentaacetate is the β-anomer of D-galactose with five acetylated hydroxyl groups. It functions as an insulin antagonist, penetrates the plasma membrane, and is hydrolyzed by intracellular esterases; it inhibits leucine-induced insulin release in Wistar rat islets. Supplied as a solid or as pre-made DMSO solutions for research use.

  • High purity (≈99.6%).
  • Molecular weight 390.34 g·mol-1.
  • Cell-permeable acetylated sugar cleaved by intracellular esterases.
  • Available as solid and ready-to-use DMSO solutions.
  • White to off-white solid appearance.
  • Store solid at 4°C under nitrogen and protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.