Learn More
Medchemexpress LLC β-D-galactose pentaacetate | 4163-60-4 | MFCD00063259 | 99.6% | 390.34 g/mol | C16H22O11 | 25 G
SDP
Catalog No. 5000303045
Encompass_Preferred
β-D-Galactose pentaacetate is the peracetylated derivative of D-galactose used as a reagent and synthetic intermediate in carbohydrate chemistry and biochemical assays. It is supplied as a white to off-white solid and is hydrolyzable by intracellular esterases; it has documented stability and storage recommendations for solids and solutions.
  • High purity for reliable experimental results.
  • Peracetylated form improves solubility in organic media.
  • Hydrolyzable by intracellular esterases for cellular studies.
  • Available as solid and solution formats for flexible use.
  • Stability data provided to support proper storage and handling.
Catalog No. 50-003-03045 Supplier Medchemexpress LLC Supplier No. 25GHYW039923
β-D-Galactose pentaacetate is the peracetylated derivative of D-galactose used as a reagent and synthetic intermediate in carbohydrate chemistry and biochemical assays. It is supplied as a white to off-white solid and is hydrolyzable by intracellular esterases; it has documented stability and storage recommendations for solids and solutions.
  • High purity for reliable experimental results.
  • Peracetylated form improves solubility in organic media.
  • Hydrolyzable by intracellular esterases for cellular studies.
  • Available as solid and solution formats for flexible use.
  • Stability data provided to support proper storage and handling.