Learn More
Cayman Chemical D-Galacturonic Acid (hydrate), Purity: ≥95%, CAS: 91510-62-2, 10 g
SDP
Catalog No. 502949955
Encompass_Preferred
A monosaccharide; a major component of pectin; increases body weight and reduces intestinal mucosal permeability in a rat model of iodoacetamide-induced functional dyspepsia at 200, 400, and 800 mg/kg
Catalog No. 50-294-9955 Supplier Cayman Chemical Supplier No. 3441610 G
A monosaccharide; a major component of pectin; increases body weight and reduces intestinal mucosal permeability in a rat model of iodoacetamide-induced functional dyspepsia at 200, 400, and 800 mg/kg